Assay Details
| Gene Symbol: Cited4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019563.2 | NP_062509.1 | 1 | 484 | 82 | |
| GenBank mRNA | AF143369.1 | - | 1 | 454 | 82 | |
| AK012136.1 | - | 1 | 476 | 82 | ||
| AK154030.1 | - | 1 | 475 | 82 | ||
| BC132360.1 | - | 1 | 431 | 40130637 | 82 | |
| BC132362.1 | - | 1 | 431 | 40130639 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cited4
Genomic Map