Assay Details
| Gene Symbol: Nudt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290181.1 | NP_001277110.1 | 1 - 2 | 1866 | 61 | |
| GenBank mRNA | AK004924.1 | - | 1 - 2 | 1822 | 61 | |
More Information
Set Membership: |
|

Nudt7
Genomic Map