Assay Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025382.6 | NP_079658.2 | 8 - 9 | 1722 | 63 | |
| GenBank mRNA | AK003528.1 | - | 2 - 3 | 238 | 63 | |
| AY364165.1 | - | 8 - 9 | 1507 | 63 | ||
| AY846873.1 | - | 8 - 9 | 1723 | 63 | ||
| AY846874.1 | - | 8 - 9 | 1723 | 63 | ||
| BC043306.1 | - | 5 - 6 | 1154 | 63 | ||
| BC054769.1 | - | 8 - 9 | 1615 | 6415086 | 63 | |
| BC056944.1 | - | 1 - 2 | 76 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK003528.1,BC056944.1 transcripts.
Set Membership: |
|

Tmem57
Genomic Map