Assay Details
| Gene Symbol: Asb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025758.4 | NP_080034.2 | 1 - 2 | 488 | 129 | |
| GenBank mRNA | AF425641.1 | - | 1 - 2 | 488 | 129 | |
| AK016273.1 | - | 1 - 2 | 488 | 129 | ||
| BC049572.1 | - | 1 - 2 | 493 | 6773716 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asb17
Genomic Map