Assay Details
| Gene Symbol: Clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027085.3 | NP_081361.1 | 3 - 4 | 311 | 74 | |
| GenBank mRNA | AK009020.1 | - | 3 - 4 | 313 | 74 | |
| BC060967.1 | - | 3 - 4 | 316 | 30297308 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clic3
Genomic Map