Assay Details
| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027088.2 | NP_081364.1 | 9 - 10 | 914 | 66 | |
| GenBank mRNA | AB047820.1 | - | 9 - 10 | 815 | 66 | |
| AK079374.1 | - | 9 - 10 | 930 | 66 | ||
| AK129108.1 | - | 9 - 10 | 822 | 66 | ||
| AK170576.1 | - | 9 - 10 | 915 | 66 | ||
| AK171561.1 | - | 9 - 10 | 916 | 66 | ||
| BC042489.1 | - | 9 - 10 | 865 | 66 | ||
| BC050901.1 | - | 9 - 10 | 877 | 6402443 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bap1
Genomic Map