Assay Details
| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027148.3 | NP_081424.3 | 4 - 5 | 528 | 92 | |
| GenBank mRNA | AJ505005.1 | - | 4 - 5 | 188 | 92 | |
| AK009584.1 | - | 4 - 5 | 200 | 92 | ||
| AK169719.1 | - | 2 - 3 | 131 | 92 | ||
| AK220227.1 | - | 4 - 5 | 528 | 92 | ||
| BC059089.1 | - | 4 - 5 | 557 | 6509820 | 92 | |
| BC067250.1 | - | 3 - 4 | 171 | 92 | ||
More Information
Set Membership: |
|

Exosc8
Genomic Map