Assay Details
| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163570.1 | NP_001157042.1 | 7 - 8 | 741 | 117 | |
| NM_027148.3 | NP_081424.3 | 7 - 8 | 729 | 117 | ||
| GenBank mRNA | AJ505005.1 | - | 7 - 8 | 389 | 117 | |
| AK009584.1 | - | 7 - 8 | 401 | 117 | ||
| AK169719.1 | - | 5 - 6 | 332 | 117 | ||
| AK220227.1 | - | 7 - 8 | 729 | 117 | ||
| BC059089.1 | - | 7 - 8 | 758 | 6509820 | 117 | |
| BC067250.1 | - | 6 - 7 | 372 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149454 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149451 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149460 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149455 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149452 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | AK220227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | BC004769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | BC059089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | BC067250.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | AK009584.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | AK220227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | BC004769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | BC059089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | BC067250.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P496260 | AK009584.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52856 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169020 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | AK220227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | BC004769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | BC059089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | BC067250.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | AK009584.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | AK220227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | BC004769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | BC059089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | BC067250.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P496260 | AK009584.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500806 | BC004769.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P500806 | AK047257.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22149460 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00645566 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169020 | - | - |
| Whole Mouse Genome G4122A | A_51_P500806 | BC004769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P500806 | AK047257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | AK220227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | BC004769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | BC059089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | BC067250.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | AK009584.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | AK220227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | BC004769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | BC059089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | BC067250.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496260 | AK009584.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103319 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02399779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02398685 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149454 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149455 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149452 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149451 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22149460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22149386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02762112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02390981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02390982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02399771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02399774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22149367 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10498174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498173 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498172 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428291_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420115_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498168 | - | - |
| Mouse Genome 430 2.0 Array | 1420115_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420115_at | - | - |
| Mouse Genome 430 2.0 Array | 1428291_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428291_at | - | - |
| Mouse Genome 430 2.0 Array | 1449738_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1449738_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69639_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69639_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300015122.mm.1 | NM_001163570.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015122.mm.1 | NM_001163570.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015122.mm.1 | NM_027148.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015122.mm.1 | NM_027148.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015122.mm.1 | TC0300001987.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428291_PM_at | - | - |
| Mouse Expression Set 430 | 1428291_at | - | - |
| Murine Genome U74v2 Set | 95081_at | - | - |
| Murine Genome U74v2 Set | 95081_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420115_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420115_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428291_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428291_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 621410 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 360427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000360427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2754369 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2754369 | - | - |
| mouse-6 v1.1 (Illumina) | scl22158.8.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 360427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22158.8.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2754369 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2754369 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl22158.8.1_12-S | - | - |

Exosc8
Genomic Map