Assay Details
| Gene Symbol: Gorasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027352.4 | NP_081628.3 | 8 - 9 | 1100 | 101 | |
| NR_027343.1 | 9 - 10 | 1297 | 101 | |||
| GenBank mRNA | AK002515.1 | - | 9 - 10 | 1178 | 101 | |
| AK033413.1 | - | 8 - 9 | 1034 | 101 | ||
| AK077153.1 | - | 8 - 9 | 975 | 101 | ||
| AK077683.1 | - | 8 - 9 | 1035 | 101 | ||
| AK077706.1 | - | 8 - 9 | 967 | 101 | ||
| AK088239.1 | - | 8 - 9 | 981 | 101 | ||
| AK148337.1 | - | 8 - 9 | 1034 | 101 | ||
| AK151843.1 | - | 8 - 9 | 982 | 101 | ||
| AK152475.1 | - | 8 - 9 | 982 | 101 | ||
| AK152544.1 | - | 8 - 9 | 982 | 101 | ||
| AK152603.1 | - | 8 - 9 | 1010 | 101 | ||
| AK153049.1 | - | 8 - 9 | 982 | 101 | ||
| AK153555.1 | - | 8 - 9 | 1033 | 101 | ||
| AK159469.1 | - | 8 - 9 | 1032 | 101 | ||
| AK168928.1 | - | 9 - 10 | 1230 | 101 | ||
| AK169382.1 | - | 8 - 9 | 982 | 101 | ||
| AK179912.1 | - | 1 - 2 | 64 | 101 | ||
| AK207289.1 | - | 2 - 3 | 195 | 101 | ||
| AK216502.1 | - | 3 - 4 | 317 | 101 | ||
| BC005600.1 | - | 8 - 9 | 973 | 3482776 | 101 | |
| BC007468.1 | - | 1 - 2 | 672 | 101 | ||
| BC016455.1 | - | 7 - 8 | 825 | 4234343 | 101 | |
| BC024982.1 | - | 8 - 9 | 986 | 101 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC007468.1 transcripts.
Set Membership: |
|

Gorasp2
Genomic Map