Assay Details
| Gene Symbol: Sugp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027481.2 | NP_081757.1 | 11 - 12 | 1866 | 88 | |
| GenBank mRNA | AF521129.1 | - | 11 - 12 | 1866 | 88 | |
| AK017736.1 | - | 11 - 12 | 1634 | 88 | ||
| AK156508.1 | - | 11 - 12 | 1643 | 88 | ||
| BC027188.1 | - | 11 - 12 | 1642 | 4035499 | 88 | |
| BC120919.1 | - | 11 - 12 | 1756 | 40093189 | 88 | |
| BC120920.1 | - | 11 - 12 | 1756 | 40093190 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sugp1
Genomic Map