Assay Details
| Gene Symbol: Arhgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289686.1 | NP_001276615.1 | 9 - 10 | 877 | 68 | |
| NM_001289687.1 | NP_001276616.1 | 6 - 7 | 748 | 68 | ||
| NM_001289688.1 | NP_001276617.1 | 6 - 7 | 922 | 68 | ||
| NM_027871.2 | NP_082147.1 | 7 - 8 | 722 | 68 | ||
| GenBank mRNA | AK004600.2 | - | 7 - 8 | 693 | 68 | |
| AK029872.1 | - | 10 - 11 | 907 | 68 | ||
| BC005517.1 | - | 6 - 7 | 719 | 3494309 | 68 | |
| BC012262.1 | - | 6 - 7 | 700 | 4192378 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef3
Genomic Map