Assay Details
| Gene Symbol: Shroom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290789.1 | NP_001277718.1 | 1 - 2 | 139 | 79 | |
| NM_027917.3 | NP_082193.2 | 1 - 2 | 139 | 79 | ||
| GenBank mRNA | AK004934.1 | - | 1 - 2 | 66 | 79 | |
| AK035462.1 | - | 1 - 2 | 99 | 79 | ||
| AK085693.1 | - | 1 - 2 | 95 | 79 | ||
| AK134332.1 | - | 1 - 2 | 67 | 79 | ||
| BC005761.1 | - | 1 - 2 | 114 | 3601742 | 79 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC005761.1,NM_001290789.1,NM_027917.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804896 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804861 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804920 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804919 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804856 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087315 | AK134332.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087315 | AK035462.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087315 | NM_027917.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175194 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140506 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105855 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087315 | AK134332.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087315 | AK035462.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087315 | NM_027917.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804874 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105855 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07345400 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140506 | - | - |
| Whole Mouse Genome G4122A | A_51_P444005 | BC031598.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444005 | AK134332.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444005 | BC005761.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444005 | AK085693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444005 | AK004934.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103248 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140506 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105855 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373865 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09184194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09742640 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09742642 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09184196 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09742636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373872 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09184197 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09742639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804896 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804919 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804861 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804920 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09039251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09684047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09684032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09742630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09135587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08997270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09184194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09684039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09626087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09373869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08997279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09546672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09465791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09480550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09050706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09373876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804881 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10376014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376012 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376009 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456371_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376007 | - | - |
| Mouse Genome 430 2.0 Array | 1456371_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456371_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71774_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71774_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100004764.mm.1 | NM_027917.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004764.mm.1 | TC1100000605.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004761.mm.1 | NM_027917.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004761.mm.1 | TC1100000605.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004762.mm.1 | NM_027917.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004762.mm.1 | TC1100000605.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004763.mm.1 | NM_027917.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004763.mm.1 | TC1100000605.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004758.mm.1 | NM_027917.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004758.mm.1 | TC1100000605.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456371_PM_a_at | - | - |
| Mouse Expression Set 430 | 1456371_a_at | - | - |
| Murine Genome U74v2 Set | 166928_at | - | - |
| Murine Genome U74v2 Set | 166928_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456371_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456371_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456371_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456371_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 337121 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4780037 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251379 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251379 | - | - |
| mouse-6 v1.1 (Illumina) | scl071774.7_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl071774.7_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2632839 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2632839 | - | - |

Shroom1
Genomic Map