Assay Details
| Gene Symbol: Rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028009.3 | NP_082285.1 | 4 - 5 | 510 | 56 | |
| GenBank mRNA | AK009926.1 | - | 3 - 4 | 342 | 56 | |
| AK049334.1 | - | 4 - 5 | 511 | 56 | ||
| BC018186.1 | - | 4 - 5 | 471 | 4160116 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpusd1
Genomic Map