Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 2 - 3 | 310 | 104 | |
| GenBank mRNA | AF319547.1 | - | 2 - 3 | 670 | 104 | |
| AK028274.1 | - | 2 - 3 | 324 | 104 | ||
| AK031534.1 | - | 2 - 3 | 320 | 104 | ||
| AK031679.1 | - | 2 - 3 | 328 | 104 | ||
| AK147715.1 | - | 2 - 3 | 325 | 104 | ||
| AK152989.1 | - | 2 - 3 | 324 | 103 | ||
| AK155138.1 | - | 2 - 3 | 322 | 104 | ||
| AK167603.1 | - | 2 - 3 | 324 | 104 | ||
| AK168455.1 | - | 2 - 3 | 324 | 104 | ||
| AK169255.1 | - | 2 - 3 | 322 | 104 | ||
| AK170967.1 | - | 2 - 3 | 322 | 104 | ||
| BC043699.1 | - | 2 - 3 | 310 | 5344158 | 104 | |
| BC062920.1 | - | 2 - 3 | 313 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map