Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 7 - 8 | 751 | 62 | |
| GenBank mRNA | AF319547.1 | - | 7 - 8 | 1111 | 62 | |
| AK010484.1 | - | 4 - 5 | 331 | 62 | ||
| AK028274.1 | - | 7 - 8 | 765 | 62 | ||
| AK031534.1 | - | 7 - 8 | 761 | 62 | ||
| AK031679.1 | - | 7 - 8 | 769 | 62 | ||
| AK147715.1 | - | 7 - 8 | 766 | 62 | ||
| AK152989.1 | - | 7 - 8 | 767 | 62 | ||
| AK155138.1 | - | 7 - 8 | 763 | 62 | ||
| AK167603.1 | - | 7 - 8 | 765 | 62 | ||
| AK168455.1 | - | 7 - 8 | 765 | 62 | ||
| AK169255.1 | - | 7 - 8 | 764 | 62 | ||
| AK170967.1 | - | 7 - 8 | 763 | 62 | ||
| AK212480.1 | - | 3 - 4 | 178 | 62 | ||
| BC043699.1 | - | 7 - 8 | 751 | 5344158 | 62 | |
| BC055026.1 | - | 3 - 4 | 194 | 62 | ||
| BC062920.1 | - | 7 - 8 | 754 | 62 | ||
| BC071251.1 | - | 1 - 2 | 48 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map