Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 11 - 12 | 1286 | 75 | |
| GenBank mRNA | AF319547.1 | - | 11 - 12 | 1646 | 75 | |
| AK010484.1 | - | 8 - 9 | 866 | 75 | ||
| AK028274.1 | - | 11 - 12 | 1300 | 75 | ||
| AK031534.1 | - | 11 - 12 | 1296 | 75 | ||
| AK031679.1 | - | 11 - 12 | 1304 | 75 | ||
| AK147715.1 | - | 11 - 12 | 1301 | 75 | ||
| AK152989.1 | - | 11 - 12 | 1302 | 75 | ||
| AK155138.1 | - | 11 - 12 | 1298 | 75 | ||
| AK167603.1 | - | 11 - 12 | 1300 | 75 | ||
| AK168455.1 | - | 11 - 12 | 1300 | 75 | ||
| AK169255.1 | - | 11 - 12 | 1299 | 75 | ||
| AK170967.1 | - | 11 - 12 | 1298 | 75 | ||
| BC021900.1 | - | 2 - 3 | 291 | 75 | ||
| BC043699.1 | - | 11 - 12 | 1286 | 5344158 | 75 | |
| BC055026.1 | - | 7 - 8 | 729 | 75 | ||
| BC062920.1 | - | 11 - 12 | 1289 | 75 | ||
| BC071251.1 | - | 5 - 6 | 583 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map