Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 12 - 13 | 1419 | 97 | |
| GenBank mRNA | AF319547.1 | - | 12 - 13 | 1779 | 97 | |
| AK010484.1 | - | 9 - 10 | 999 | 97 | ||
| AK028274.1 | - | 12 - 13 | 1433 | 97 | ||
| AK031534.1 | - | 12 - 13 | 1429 | 97 | ||
| AK031679.1 | - | 12 - 13 | 1437 | 97 | ||
| AK147715.1 | - | 12 - 13 | 1434 | 97 | ||
| AK152989.1 | - | 12 - 13 | 1435 | 97 | ||
| AK155138.1 | - | 12 - 13 | 1431 | 97 | ||
| AK167603.1 | - | 12 - 13 | 1433 | 97 | ||
| AK168455.1 | - | 12 - 13 | 1433 | 97 | ||
| AK169255.1 | - | 12 - 13 | 1432 | 97 | ||
| AK170967.1 | - | 12 - 13 | 1431 | 97 | ||
| BC021900.1 | - | 3 - 4 | 424 | 97 | ||
| BC043699.1 | - | 12 - 13 | 1419 | 5344158 | 97 | |
| BC055026.1 | - | 8 - 9 | 862 | 97 | ||
| BC062920.1 | - | 12 - 13 | 1422 | 97 | ||
| BC071251.1 | - | 6 - 7 | 716 | 97 | ||
More Information
Set Membership: |
|

Ddx54
Genomic Map