Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 13 - 14 | 1653 | 101 | |
| GenBank mRNA | AF319547.1 | - | 13 - 14 | 2013 | 101 | |
| AK028274.1 | - | 13 - 14 | 1667 | 101 | ||
| AK031534.1 | - | 13 - 14 | 1663 | 101 | ||
| AK031679.1 | - | 13 - 14 | 1671 | 101 | ||
| AK152989.1 | - | 13 - 14 | 1669 | 101 | ||
| AK155138.1 | - | 13 - 14 | 1665 | 101 | ||
| AK167603.1 | - | 13 - 14 | 1667 | 101 | ||
| AK168455.1 | - | 13 - 14 | 1667 | 101 | ||
| AK169255.1 | - | 13 - 14 | 1666 | 101 | ||
| AK170967.1 | - | 13 - 14 | 1665 | 101 | ||
| AK184886.1 | - | 1 - 2 | 117 | 101 | ||
| BC021900.1 | - | 4 - 5 | 658 | 101 | ||
| BC043699.1 | - | 13 - 14 | 1653 | 5344158 | 101 | |
| BC055026.1 | - | 9 - 10 | 1096 | 101 | ||
| BC062920.1 | - | 13 - 14 | 1656 | 101 | ||
| BC071251.1 | - | 7 - 8 | 950 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map