Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 15 - 16 | 1924 | 61 | |
| GenBank mRNA | AF319547.1 | - | 15 - 16 | 2284 | 61 | |
| AK028274.1 | - | 15 - 16 | 1939 | 61 | ||
| AK031534.1 | - | 15 - 16 | 1934 | 61 | ||
| AK031679.1 | - | 15 - 16 | 1942 | 61 | ||
| AK147715.1 | - | 15 - 16 | 1940 | 61 | ||
| AK152989.1 | - | 15 - 16 | 1940 | 61 | ||
| AK155138.1 | - | 15 - 16 | 1936 | 61 | ||
| AK167603.1 | - | 15 - 16 | 1938 | 61 | ||
| AK168455.1 | - | 15 - 16 | 1938 | 61 | ||
| AK169255.1 | - | 15 - 16 | 1937 | 61 | ||
| AK170967.1 | - | 15 - 16 | 1936 | 61 | ||
| BC021900.1 | - | 6 - 7 | 929 | 61 | ||
| BC043699.1 | - | 15 - 16 | 1924 | 5344158 | 61 | |
| BC055026.1 | - | 11 - 12 | 1367 | 61 | ||
| BC071251.1 | - | 9 - 10 | 1221 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map