Assay Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 18 - 19 | 2280 | 67 | |
| GenBank mRNA | AF319547.1 | - | 18 - 19 | 2640 | 67 | |
| AK010484.1 | - | 15 - 16 | 1861 | 67 | ||
| AK028274.1 | - | 18 - 19 | 2295 | 67 | ||
| AK031534.1 | - | 18 - 19 | 2290 | 67 | ||
| AK031679.1 | - | 18 - 19 | 2298 | 67 | ||
| AK147715.1 | - | 18 - 19 | 2296 | 67 | ||
| AK152989.1 | - | 18 - 19 | 2296 | 67 | ||
| AK155138.1 | - | 18 - 19 | 2292 | 67 | ||
| AK167603.1 | - | 18 - 19 | 2294 | 67 | ||
| AK168455.1 | - | 18 - 19 | 2294 | 67 | ||
| AK169255.1 | - | 18 - 19 | 2293 | 67 | ||
| AK170967.1 | - | 18 - 19 | 2292 | 67 | ||
| BC021900.1 | - | 9 - 10 | 1285 | 67 | ||
| BC043699.1 | - | 18 - 19 | 2280 | 5344158 | 67 | |
| BC055026.1 | - | 14 - 15 | 1723 | 67 | ||
| BC062920.1 | - | 16 - 17 | 3616 | 67 | ||
| BC071251.1 | - | 12 - 13 | 1577 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx54
Genomic Map