Assay Details
| Gene Symbol: Brf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028193.3 | NP_082469.2 | 17 - 18 | 2265 | 70 | |
| GenBank mRNA | AK010890.1 | - | 4 - 5 | 514 | 70 | |
| BC037224.1 | - | 17 - 18 | 2048 | 3257364 | 70 | |
| BC058112.1 | - | 7 - 8 | 861 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase III promoter
regulation of mRNA stability
positive regulation of transcription from RNA polymerase III promoter
molecular_function
zinc ion binding
TBP-class protein binding
metal ion binding
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase III promoter
regulation of mRNA stability
positive regulation of transcription from RNA polymerase III promoter
molecular_function
zinc ion binding
TBP-class protein binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | BC037224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | BC058112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | NM_028193.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | AK010890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | BC037224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | BC058112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | NM_028193.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P592111 | AK010890.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129748 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735320 | AK016905.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735320 | AK034243.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735320 | AK083617.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735320 | AK084829.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735320 | BC118051.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735320 | BC119793.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735320 | NM_027695.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907668 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89321 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31880 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108430 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121870 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105891 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147388 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | BC037224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | BC058112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | NM_028193.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | AK010890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | BC037224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | BC058112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | NM_028193.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P592111 | AK010890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | BC037224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | BC058112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | NM_028193.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | AK010890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | BC037224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | BC058112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | NM_028193.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592111 | AK010890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P867427 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111588 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10226007 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10223626 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10223620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10223619 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10223625 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10402844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402856 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402859 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402855 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402854 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402863 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402860 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402857 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402861 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402849 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453209_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398934 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402841 | - | - |
| Mouse Genome 430 2.0 Array | 1453209_at | - | - |
| Mouse Genome 430 2.0 Array | 1453209_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72308_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72308_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200017008.mm.1 | NM_028193.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200017008.mm.1 | NM_028193.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200017008.mm.1 | TC1200002448.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200017009.mm.1 | NM_028193.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200017009.mm.1 | NM_028193.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200017009.mm.1 | TC1200002448.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453209_at | - | - |
| Mouse Expression Set 430 | 1453209_PM_at | - | - |
| Murine Genome U74v2 Set | 166331_at | - | - |
| Murine Genome U74v2 Set | 166331_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453209_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453209_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 515067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2100047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220096 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220096 | - | - |
| mouse-6 v1.1 (Illumina) | scl42009.20.1_59-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100047 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42009.20.1_59-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220096 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220096 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl42009.20.1_59-S | - | - |

Brf1
Genomic Map