Assay Details
| Gene Symbol: Tdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028354.4 | NP_082630.2 | 7 - 8 | 932 | 70 | |
| GenBank mRNA | AK013427.1 | - | 6 - 7 | 407 | 70 | |
| AK014855.1 | - | 7 - 8 | 912 | 70 | ||
| AK033868.1 | - | 7 - 8 | 930 | 70 | ||
| BC065162.1 | - | 6 - 7 | 902 | 6402921 | 70 | |
More Information
Set Membership: |
|

Tdp1
Genomic Map