Assay Details
| Gene Symbol: Phykpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028398.2 | NP_082674.1 | 3 - 4 | 400 | 64 | |
| GenBank mRNA | AK013489.1 | - | 3 - 4 | 354 | 64 | |
| AK085984.1 | - | 3 - 4 | 351 | 64 | ||
| AK133235.1 | - | 347 | 64 | |||
| AK156866.1 | - | 3 - 4 | 401 | 64 | ||
| AK160557.1 | - | 3 - 4 | 369 | 64 | ||
| AK217003.1 | - | 1 - 2 | 85 | 64 | ||
| BC024461.1 | - | 3 - 4 | 354 | 4975449 | 64 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK087703.1 | - | 347 | Chr. - - [-] on Build | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phykpl
Genomic Map