Assay Details
| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028447.3 | NP_082723.2 | 5 - 6 | 933 | 103 | |
| GenBank mRNA | AK143537.1 | - | 5 - 6 | 925 | 103 | |
| BC014817.1 | - | 5 - 6 | 849 | 3709357 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prrc1
Genomic Map