Assay Details
| Gene Symbol: Pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028994.2 | NP_083270.1 | 2 - 3 | 493 | 73 | |
| GenBank mRNA | AK030501.1 | - | 2 - 3 | 494 | 73 | |
| AK031612.1 | - | 2 - 3 | 338 | 73 | ||
| AK034927.1 | - | 2 - 3 | 372 | 73 | ||
| AK147972.1 | - | 2 - 3 | 347 | 73 | ||
| AK165094.1 | - | 2 - 3 | 371 | 73 | ||
| AK170242.1 | - | 2 - 3 | 347 | 73 | ||
| BC010318.1 | - | 2 - 3 | 356 | 3596227 | 73 | |
| BC011126.1 | - | 2 - 3 | 353 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pck2
Genomic Map