Assay Details
| Gene Symbol: Pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028994.2 | NP_083270.1 | 3 - 4 | 678 | 70 | |
| GenBank mRNA | AK030501.1 | - | 3 - 4 | 679 | 70 | |
| AK031612.1 | - | 3 - 4 | 523 | 70 | ||
| AK034927.1 | - | 3 - 4 | 557 | 70 | ||
| AK147972.1 | - | 3 - 4 | 532 | 70 | ||
| AK165094.1 | - | 3 - 4 | 556 | 70 | ||
| AK170242.1 | - | 3 - 4 | 532 | 70 | ||
| BC010318.1 | - | 3 - 4 | 541 | 3596227 | 70 | |
| BC011126.1 | - | 3 - 4 | 538 | 70 | ||
| BC023437.1 | - | 1 - 2 | 146 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pck2
Genomic Map