Assay Details
| Gene Symbol: Nln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029447.2 | NP_083723.1 | 7 - 8 | 1110 | 70 | |
| GenBank mRNA | AK079090.1 | - | 7 - 8 | 1111 | 70 | |
| AK138399.1 | - | 7 - 8 | 1083 | 70 | ||
| AK146364.1 | - | 7 - 8 | 1059 | 70 | ||
| AK170508.1 | - | 7 - 8 | 1111 | 70 | ||
| AK171432.1 | - | 7 - 8 | 1111 | 70 | ||
| BC016224.1 | - | 7 - 8 | 1083 | 4486941 | 70 | |
More Information
Set Membership: |
|

Nln
Genomic Map