Assay Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252437.1 | NP_001239366.1 | 11 - 12 | 1352 | 68 | |
| NM_029657.4 | NP_083933.1 | 11 - 12 | 1349 | 68 | ||
| GenBank mRNA | AK011747.2 | - | 11 - 12 | 1142 | 68 | |
| AK088533.1 | - | 11 - 12 | 1132 | 68 | ||
| AK129161.1 | - | 8 - 9 | 760 | 68 | ||
| AK150176.1 | - | 12 - 13 | 1185 | 68 | ||
| AK153407.1 | - | 11 - 12 | 1173 | 68 | ||
| AK211037.1 | - | 1 - 2 | 76 | 68 | ||
| BC046830.1 | - | 11 - 12 | 1198 | 6811606 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgrn1
Genomic Map