Assay Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001326.1 | NP_001001326.1 | 15 - 16 | 3008 | 76 | |
| NM_029811.2 | NP_084087.2 | 14 - 15 | 1757 | 76 | ||
| NR_037617.1 | 15 - 16 | 3189 | 76 | |||
| GenBank mRNA | AK008100.1 | - | 7 - 8 | 831 | 76 | |
| BC030391.1 | - | 11 - 12 | 1483 | 4950637 | 76 | |
| BC032266.1 | - | 7 - 8 | 903 | 76 | ||
| BC051034.1 | - | 15 - 16 | 3189 | 5345299 | 76 | |
More Information
Set Membership: |
|

St5
Genomic Map