Assay Details
| Gene Symbol: Car7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301164.1 | NP_001288093.1 | 3 - 4 | 841 | 65 | |
| NM_001301165.1 | NP_001288094.1 | 3 - 4 | 409 | 65 | ||
| NM_053070.3 | NP_444300.1 | 3 - 4 | 436 | 65 | ||
| GenBank mRNA | AF291660.1 | - | 2 - 3 | 331 | 65 | |
| AK144275.1 | - | 3 - 4 | 410 | 65 | ||
| AY075021.1 | - | 3 - 4 | 437 | 65 | ||
| BC038270.1 | - | 3 - 4 | 816 | 65 | ||
| BC094913.1 | - | 3 - 4 | 424 | 30016726 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car7
Genomic Map