Assay Details
| Gene Symbol: Car7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301164.1 | NP_001288093.1 | 5 - 6 | 1000 | 61 | |
| NM_001301165.1 | NP_001288094.1 | 5 - 6 | 568 | 61 | ||
| NM_053070.3 | NP_444300.1 | 5 - 6 | 595 | 61 | ||
| GenBank mRNA | AF291660.1 | - | 4 - 5 | 490 | 61 | |
| AK090390.1 | - | 5 - 6 | 599 | 61 | ||
| AK144275.1 | - | 5 - 6 | 569 | 61 | ||
| AY075021.1 | - | 5 - 6 | 596 | 61 | ||
| BC038270.1 | - | 5 - 6 | 975 | 61 | ||
| BC094913.1 | - | 5 - 6 | 583 | 30016726 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car7
Genomic Map