Assay Details
| Gene Symbol: Fam64a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144526.3 | NP_653109.2 | 2 - 3 | 352 | 74 | |
| GenBank mRNA | AK011345.1 | - | 2 - 3 | 351 | 74 | |
| AK049710.1 | - | 2 - 3 | 353 | 74 | ||
| AK078429.1 | - | 2 - 3 | 353 | 74 | ||
| BC029115.1 | - | 2 - 3 | 338 | 3983148 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam64a
Genomic Map