Assay Details
| Gene Symbol: Klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145479.4 | NP_663454.3 | 5 - 6 | 1428 | 88 | |
| GenBank mRNA | AK143514.1 | - | 5 - 6 | 1373 | 88 | |
| BC005800.1 | - | 5 - 6 | 1406 | 2812589 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl22
Genomic Map