Assay Details
| Gene Symbol: Slc24a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172152.2 | NP_742164.1 | 15 - 16 | 1622 | 80 | |
| GenBank mRNA | AK044239.1 | - | 15 - 16 | 2254 | 80 | |
| AY156046.1 | - | 15 - 16 | 1622 | 80 | ||
| BC137945.1 | - | 15 - 16 | 1593 | 8860964 | 80 | |
| BC171939.1 | - | 15 - 16 | 1593 | 9054225 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc24a4
Genomic Map