Assay Details
| Gene Symbol: Pde8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170669.1 | NP_001164140.1 | 4 - 5 | 889 | 60 | |
| NM_172263.2 | NP_758467.2 | 4 - 5 | 481 | 60 | ||
| GenBank mRNA | AK036090.1 | - | 5 - 6 | 809 | 60 | |
| AK043680.1 | - | 4 - 5 | 882 | 60 | ||
| AK045991.1 | - | 4 - 5 | 890 | 60 | ||
| AK046298.1 | - | 4 - 5 | 905 | 60 | ||
| AK083166.1 | - | 4 - 5 | 527 | 60 | ||
| BC032286.1 | - | 4 - 5 | 493 | 5367055 | 60 | |
| CD806392.1 | - | 4 - 5 | 454 | 60 | ||
| EF644173.1 | - | 4 - 5 | 343 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde8b
Genomic Map