Assay Details
| Gene Symbol: Pde8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172263.2 | NP_758467.2 | 8 - 9 | 846 | 57 | |
| GenBank mRNA | BC032286.1 | - | 8 - 9 | 858 | 5367055 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|

Pde8b
Genomic Map