Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140537.1 | - | 1 - 2 | 1666 | 97 | |
| BC039956.1 | - | 16 - 17 | 2537 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210018M11Rik| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140537.1 | - | 1 - 2 | 1666 | 97 | |
| BC039956.1 | - | 16 - 17 | 2537 | 97 | ||
Set Membership: |
|