Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172280.2 | NP_758484.2 | 17 - 18 | 2698 | 60 | |
| GenBank mRNA | AK008751.1 | - | 4 - 5 | 680 | 60 | |
| BC020109.1 | - | 2 - 3 | 199 | 60 | ||
| BC039956.1 | - | 18 - 19 | 2730 | 60 | ||
| BC066050.1 | - | 13 - 14 | 2237 | 60 | ||
| BC089304.1 | - | 17 - 18 | 2698 | 6854147 | 60 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC020109.1 transcripts.
Set Membership: |
|

2210018M11Rik
Genomic Map