Assay Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 2 - 3 | 889 | 88 | |
GenBank mRNA | AK033499.1 | - | 2 - 3 | 650 | 88 | |
AK046065.1 | - | 2 - 3 | 679 | 88 | ||
AK049974.1 | - | 2 - 3 | 648 | 88 | ||
AK083321.1 | - | 2 - 3 | 713 | 88 | ||
AK088866.1 | - | 2 - 3 | 673 | 88 | ||
AK148252.1 | - | 2 - 3 | 692 | 88 | ||
BC039955.1 | - | 2 - 3 | 635 | 4236297 | 88 | |
BC053487.1 | - | 2 - 3 | 664 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
