Assay Details
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172338.2 | NP_758841.1 | 3 - 4 | 302 | 74 | |
GenBank mRNA | AK028692.1 | - | 3 - 4 | 307 | 74 | |
AK051048.1 | - | 3 - 4 | 303 | 74 | ||
AK122407.1 | - | 549 | 74 | |||
AK154946.1 | - | 323 | 74 | |||
AK164023.1 | - | 3 - 4 | 318 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
