Assay Details
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172338.2 | NP_758841.1 | 5 - 6 | 828 | 78 | |
GenBank mRNA | AK028692.1 | - | 5 - 6 | 833 | 78 | |
AK051048.1 | - | 5 - 6 | 829 | 78 | ||
AK122407.1 | - | 1075 | 78 | |||
AK154946.1 | - | 849 | 78 | |||
AK164023.1 | - | 5 - 6 | 844 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
