Assay Details
| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300827.1 | NP_001287756.1 | 3 - 4 | 614 | 80 | |
| NM_172380.4 | NP_759012.1 | 3 - 4 | 411 | 80 | ||
| GenBank mRNA | AK031608.1 | - | 3 - 4 | 350 | 80 | |
| AK035948.1 | - | 3 - 4 | 359 | 80 | ||
| AK036224.1 | - | 3 - 4 | 341 | 80 | ||
| AK041321.1 | - | 3 - 4 | 349 | 80 | ||
| AK045278.1 | - | 3 - 4 | 559 | 80 | ||
| AK079886.1 | - | 3 - 4 | 380 | 80 | ||
| AK167641.1 | - | 3 - 4 | 581 | 80 | ||
| AK204867.1 | - | 1 - 2 | 40 | 80 | ||
| BC026809.1 | - | 3 - 4 | 362 | 4205469 | 80 | |
| CB249259.1 | - | 3 - 4 | 412 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Poglut1
Genomic Map