Assay Details
| Gene Symbol: Slc25a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172436.3 | NP_766024.1 | 10 - 11 | 1044 | 86 | |
| GenBank mRNA | AK019150.1 | - | 10 - 11 | 1047 | 86 | |
| AK030595.1 | - | 10 - 11 | 1074 | 86 | ||
| AK083156.1 | - | 10 - 11 | 1045 | 86 | ||
| AK144958.1 | - | 10 - 11 | 1078 | 86 | ||
| AK147948.1 | - | 10 - 11 | 1059 | 86 | ||
| AK205888.1 | - | 2 - 3 | 134 | 86 | ||
| BC013845.1 | - | 10 - 11 | 1023 | 86 | ||
| BC024952.1 | - | 10 - 11 | 1237 | 86 | ||
| BC060505.1 | - | 10 - 11 | 1031 | 6311836 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a12
Genomic Map