Assay Details
| Gene Symbol: Slc25a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172436.3 | NP_766024.1 | 11 - 12 | 1206 | 65 | |
| GenBank mRNA | AK019150.1 | - | 11 - 12 | 1209 | 65 | |
| AK030595.1 | - | 11 - 12 | 1236 | 65 | ||
| AK083156.1 | - | 11 - 12 | 1207 | 65 | ||
| AK144958.1 | - | 11 - 12 | 1240 | 65 | ||
| AK217682.1 | - | 1 - 2 | 120 | 65 | ||
| BC013845.1 | - | 11 - 12 | 1185 | 65 | ||
| BC024952.1 | - | 11 - 12 | 1399 | 65 | ||
| BC060505.1 | - | 11 - 12 | 1193 | 6311836 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a12
Genomic Map