Assay Details
| Gene Symbol: Galnt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161767.1 | NP_001155239.1 | 7 - 8 | 1556 | 142 | |
| NM_001161768.1 | NP_001155240.1 | 7 - 8 | 1553 | 142 | ||
| NM_172451.3 | NP_766039.2 | 7 - 8 | 1552 | 142 | ||
| GenBank mRNA | AJ133523.1 | - | 5 - 6 | 1168 | 142 | |
| AK028506.1 | - | 7 - 8 | 1552 | 142 | ||
| AK049222.1 | - | 7 - 8 | 1555 | 142 | ||
| AK155008.1 | - | 7 - 8 | 1559 | 142 | ||
| AK159721.1 | - | 7 - 8 | 1534 | 142 | ||
| BC119324.1 | - | 5 - 6 | 1205 | 8734073 | 142 | |
| BC119326.1 | - | 5 - 6 | 1205 | 8734075 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt6
Genomic Map