Assay Details
| Gene Symbol: Panx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172454.2 | NP_766042.2 | 3 - 4 | 708 | 127 | |
| GenBank mRNA | AK028288.1 | - | 3 - 4 | 675 | 127 | |
| AK036933.1 | - | 3 - 4 | 664 | 127 | ||
| BC052822.1 | - | 3 - 4 | 676 | 30058169 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P27190 | AK036933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P27190 | NM_172454.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P27190 | AK028288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P27190 | AK036933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P27190 | NM_172454.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P27190 | AK028288.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243601 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489618 | AK017229.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489618 | AK138939.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489618 | AK214115.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489618 | BC023339.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489618 | NM_153196.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 879667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101248 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173568 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P27190 | AK036933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P27190 | NM_172454.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P27190 | AK028288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P27190 | AK036933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P27190 | NM_172454.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P27190 | AK028288.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431968 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173568 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06754156 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06754149 | - | - |
| Whole Mouse Genome G4122A | A_52_P27190 | AK036933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27190 | NM_172454.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27190 | AK028288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27190 | AK036933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27190 | NM_172454.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27190 | AK028288.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111768 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537465 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537456 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537459 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537464 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07537472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07905580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07540854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08014644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08129709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07624584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07964983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07905372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07537460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07905577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07624572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08067524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07501459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432031 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592359 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592358 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592360 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456073_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454387_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432860_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592355 | - | - |
| Mouse Genome 430 2.0 Array | 1456073_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1432860_at | - | - |
| Mouse Genome 430 2.0 Array | 1432860_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456073_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1454387_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454387_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208098_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208098_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900017156.mm.1 | NM_172454.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017156.mm.1 | NM_172454.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017156.mm.1 | TC0900002055.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900017155.mm.1 | NM_172454.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017155.mm.1 | NM_172454.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017155.mm.1 | TC0900002055.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456073_PM_s_at | - | - |
| Mouse Expression Set 430 | 1432860_PM_at | - | - |
| Mouse Expression Set 430 | 1454387_PM_at | - | - |
| Mouse Expression Set 430 | 1454387_at | - | - |
| Mouse Expression Set 430 | 1456073_s_at | - | - |
| Mouse Expression Set 430 | 1432860_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456073_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454387_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454387_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432860_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456073_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432860_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 502263 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6180072 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6180072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721052 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2721052 | - | - |
| mouse-6 v1.1 (Illumina) | scl36021.4.1_26-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180072 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36021.4.1_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180072 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2721052 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2721052 | - | - |

Panx3
Genomic Map