Assay Details
| Gene Symbol: Nek11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172461.3 | NP_766049.2 | 5 - 6 | 748 | 113 | |
| GenBank mRNA | AK030042.1 | - | 5 - 6 | 707 | 113 | |
| AK043543.1 | - | 5 - 6 | 987 | 113 | ||
| AK054237.1 | - | 5 - 6 | 642 | 113 | ||
| BC119588.1 | - | 4 - 5 | 582 | 40058889 | 113 | |
| BC119589.1 | - | 4 - 5 | 582 | 40058890 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek11
Genomic Map