Assay Details
| Gene Symbol: Zdhhc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172465.4 | NP_766053.1 | 2 - 3 | 517 | 85 | |
| GenBank mRNA | AK032233.1 | - | 2 - 3 | 499 | 85 | |
| AK035745.1 | - | 1 - 2 | 1940 | 85 | ||
| AK079248.1 | - | 2 - 3 | 484 | 85 | ||
| BC042618.1 | - | 2 - 3 | 376 | 4950217 | 85 | |
| BC090832.1 | - | 2 - 3 | 335 | 6401763 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc9
Genomic Map