Assay Details
| Gene Symbol: Zdhhc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172465.4 | NP_766053.1 | 7 - 8 | 1120 | 66 | |
| GenBank mRNA | AK032233.1 | - | 7 - 8 | 1102 | 66 | |
| BC042618.1 | - | 7 - 8 | 979 | 4950217 | 66 | |
| BC090832.1 | - | 7 - 8 | 938 | 6401763 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc9
Genomic Map