Assay Details
| Gene Symbol: Zfp180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172483.2 | NP_766071.1 | 2 - 3 | 424 | 86 | |
| GenBank mRNA | AK083802.1 | - | 2 - 3 | 398 | 86 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK083802.1,NM_172483.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170414 | BC085472.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170414 | BC066115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170414 | BC063741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170414 | AK154182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170414 | AK145004.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936212 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618201 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677151 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99536 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136289 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180307 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170414 | BC085472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170414 | BC066115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170414 | BC063741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170414 | AK154182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170414 | AK145004.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P224818 | BC085472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P224818 | BC066115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P224818 | BC063741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P224818 | AK154182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P224818 | AY166794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157737 | BC085472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157737 | BC066115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157737 | BC063741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157737 | AK144892.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157737 | AK083802.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06345405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06103636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06345404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25003457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06380141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06329546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06329541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06460998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06314386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06329543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06329547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25003467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06144909 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550838 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428170_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550833 | - | - |
| Mouse Genome 430 2.0 Array | 1428170_at | - | - |
| Mouse Genome 430 2.0 Array | 1428170_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210135_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210135_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700002652.mm.1 | TC0700000368.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002652.mm.1 | NM_001045486.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002652.mm.1 | NM_001289634.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002652.mm.1 | NM_001289640.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002652.mm.1 | NM_001289641.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002652.mm.1 | NM_172483.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002651.mm.1 | TC0700000368.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002651.mm.1 | NM_001045486.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002651.mm.1 | NM_001289634.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002651.mm.1 | NM_001289637.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002651.mm.1 | NM_001289640.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002651.mm.1 | NM_001289641.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002651.mm.1 | NM_172483.2 | Refseq |
| Mouse Expression Set 430 | 1428170_at | - | - |
| Mouse Expression Set 430 | 1428170_PM_at | - | - |
| Murine Genome U74v2 Set | 170098_r_at | - | - |
| Murine Genome U74v2 Set | 170098_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428170_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428170_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 865203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4150592 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5720047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2803368 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2803368 | - | - |

Zfp180
Genomic Map